Search Thermo Fisher Scientific
Gene Symbol: Mapkapk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008551.1 | NP_032577.1 | 8 | 933 | ||
GenBank mRNA | AK190002.1 | - | 1 | 51 | ||
AK192111.1 | - | 1 | 51 | |||
AK198261.1 | - | 1 | 51 | |||
AK206582.1 | - | 1 | 51 | |||
BC024559.1 | - | 4 | 302 | |||
BC063064.1 | - | 8 | 933 | 5687007 | ||
X76850.1 | - | 8 | 912 |