Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Masp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003893.2 | NP_001003893.1 | 4 | 497 | ||
NM_010767.3 | NP_034897.1 | 4 | 497 | |||
GenBank mRNA | AB009459.1 | - | 4 | 487 | ||
AI529228.1 | - | 4 | 498 | |||
AJ250369.1 | - | 4 | 486 | |||
AK050052.1 | - | 4 | 474 | |||
AK149421.1 | - | 3 | 551 | |||
BC013893.1 | - | 3 | 471 | 4211293 | ||
BF233497.1 | - | 4 | 495 | |||
Y19160.1 | - | 3 | 544 | |||
Y19163.1 | - | 3 | 514 |