Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pick1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045558.1 | NP_001039023.1 | 4 | 523 | ||
NM_008837.3 | NP_032863.2 | 4 | 582 | |||
GenBank mRNA | AK050224.1 | - | 4 | 494 | ||
AK090155.1 | - | 515 | ||||
BC023088.1 | - | 4 | 489 | |||
BC048788.1 | - | 4 | 468 | 6488048 | ||
BU057774.1 | - | 4 | 485 | |||
CA468666.1 | - | 4 | 403 | |||
CD363874.1 | - | 1 | 12 | |||
CK791908.1 | - | 4 | 523 | |||
Z46720.1 | - | 3 | 271 |