Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMYD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006062.2 | NP_006053.2 | 5 | 514 | ||
XM_006711918.3 | XP_006711981.1 | 5 | 601 | |||
XM_017003163.1 | XP_016858652.1 | 4 | 2234 | |||
GenBank mRNA | AK300779.1 | - | 5 | 488 | ||
AK301809.1 | - | 5 | 507 | |||
BC073806.1 | - | 5 | 490 | 6671194 | ||
DC303248.1 | - | 5 | 514 | |||
U50383.1 | - | 5 | 467 |