Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Stmn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009133.3 | NP_033159.1 | 5 | 824 | ||
GenBank mRNA | AB007912.1 | - | 5 | 612 | ||
AF069708.1 | - | 5 | 593 | |||
AK002957.1 | - | 5 | 582 | |||
AK003203.1 | - | 5 | 582 | |||
AK003409.1 | - | 5 | 597 | |||
AK032737.1 | - | 5 | 825 | |||
AK149129.1 | - | 71 | ||||
BC057017.1 | - | 5 | 600 | 6818794 |