Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sprr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009264.2 | NP_033290.1 | 2 | 677 | ||
GenBank mRNA | AK144782.1 | - | 2 | 691 | ||
BC027534.1 | - | 2 | 677 | 1362548 | ||
X91824.1 | - | 2 | 670 |