Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stfa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082545.1 | NP_001076014.1 | 3 | 241 | ||
GenBank mRNA | BC138004.1 | - | 3 | 335 | 8861023 | |
BY780023.1 | - | 3 | 241 | |||
M92418.1 | - | 3 | 213 |