Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC4M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144904.1 | NP_001138376.1 | 4 | 714 | ||
NM_001144905.1 | NP_001138377.1 | 5 | 795 | |||
NM_001144909.1 | NP_001138381.1 | 5 | 729 | |||
NM_001144910.1 | NP_001138382.1 | 5 | 798 | |||
NM_001144911.1 | NP_001138383.1 | 3 | 783 | |||
NM_014257.4 | NP_055072.3 | 4 | 867 | |||
NR_026707.1 | 4 | 867 | ||||
NR_026708.1 | 4 | 867 | ||||
NR_026709.1 | 3 | 804 | ||||
XM_006722611.2 | XP_006722674.1 | 4 | 799 | |||
XM_006722612.3 | XP_006722675.1 | 3 | 873 | |||
XM_006722613.3 | XP_006722676.1 | 3 | 875 | |||
XM_006722614.3 | XP_006722677.1 | 2 | 791 | |||
XM_006722615.2 | XP_006722678.1 | 4 | 867 | |||
GenBank mRNA | AB015629.1 | - | 3 | 689 | ||
AF245219.2 | - | 4 | 773 | |||
AF290887.1 | - | 5 | 733 | |||
AK023750.1 | - | 2 | 588 | |||
AK292278.1 | - | 5 | 794 | |||
AK298084.1 | - | 3 | 688 | |||
AK304499.1 | - | 4 | 647 | |||
AK314704.1 | - | 5 | 728 | |||
AY042234.1 | - | 4 | 749 | |||
AY042235.1 | - | 4 | 749 | |||
AY042236.1 | - | 4 | 749 | |||
AY042239.1 | - | 3 | 686 | |||
AY343913.1 | - | 5 | 612 | |||
BC038851.1 | - | 4 | 821 | 5166966 | ||
BC110614.2 | - | 2 | 722 | 40032453 |