Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRVI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098579.2 | NP_001092049.2 | 8 | 977 | ||
NM_001100163.2 | NP_001093633.1 | 9 | 1255 | |||
NM_001100167.2 | NP_001093637.1 | 9 | 972 | |||
NM_001206881.1 | NP_001193810.1 | 7 | 1097 | |||
NM_130385.3 | NP_569056.4 | 9 | 1308 | |||
GenBank mRNA | AF081249.1 | - | 7 | 993 | ||
AF081250.1 | - | 7 | 721 | |||
AK127209.1 | - | 9 | 950 | |||
AK296336.1 | - | 9 | 1072 | |||
AK300358.1 | - | 8 | 962 | |||
BC117127.1 | - | 9 | 845 | 40125678 |