Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRVI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098579.2 | NP_001092049.2 | 10 | 1562 | ||
NM_001100163.2 | NP_001093633.1 | 11 | 1840 | |||
NM_001100167.2 | NP_001093637.1 | 11 | 1557 | |||
NM_001206880.1 | NP_001193809.1 | 10 | 1275 | |||
NM_001206881.1 | NP_001193810.1 | 9 | 1682 | |||
NM_130385.3 | NP_569056.4 | 11 | 1893 | |||
GenBank mRNA | AF081249.1 | - | 9 | 1578 | ||
AF081250.1 | - | 9 | 1306 | |||
AK127209.1 | - | 11 | 1535 | |||
AK296336.1 | - | 11 | 1657 | |||
AK300358.1 | - | 10 | 1547 | |||
AK301296.1 | - | 10 | 886 | |||
AK304702.1 | - | 10 | 909 | |||
BC117127.1 | - | 11 | 1430 | 40125678 |