Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207176.3 | NP_997059.1 | 2, 3 | 249 | ||
GenBank mRNA | AK132249.1 | - | 2, 3 | 206 | ||
AK145609.1 | - | 2, 3 | 198 | |||
AK167101.1 | - | 2, 3 | 198 | |||
AK167274.1 | - | 2, 3 | 206 | |||
BC003808.1 | - | 4, 5 | 489 | 3589585 | ||
BC010465.1 | - | 2, 3 | 205 | 3585670 | ||
BC051103.1 | - | 2, 3 | 183 | |||
CT010319.1 | - | 1, 2 | 84 |