Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpd52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025261.1 | NP_001020432.1 | 8 | 741 | ||
NM_001025262.1 | NP_001020433.1 | 6 | 672 | |||
NM_001025263.1 | NP_001020434.1 | 7 | 617 | |||
NM_001025264.1 | NP_001020435.1 | 7 | 602 | |||
NM_009412.2 | NP_033438.1 | 6 | 575 | |||
GenBank mRNA | AK005456.1 | - | 6 | 556 | ||
AK032111.1 | - | 7 | 580 | |||
AK150935.1 | - | 6 | 538 | |||
AK151471.1 | - | 6 | 536 | |||
AK151744.1 | - | 6 | 536 | |||
AK152543.1 | - | 6 | 536 | |||
AK153266.1 | - | 6 | 536 | |||
AK153292.1 | - | 6 | 546 | |||
AK161753.1 | - | 6 | 537 | |||
AK168379.1 | - | 6 | 531 | |||
AK168817.1 | - | 6 | 539 | |||
AK169751.1 | - | 6 | 568 | |||
AK170725.1 | - | 6 | 521 | |||
AK171050.1 | - | 6 | 516 | |||
AK181915.1 | - | 2 | 103 | |||
AK190065.1 | - | 1 | 19 | |||
AK198625.1 | - | 2 | 103 | |||
AK218627.1 | - | 2 | 104 | |||
AY048852.1 | - | 6 | 664 | |||
BC002036.1 | - | 6 | 555 | 3593963 | ||
BC004068.1 | - | 6 | 508 | 3590969 | ||
BC094018.1 | - | 6 | 531 | 3674995 | ||
CA480795.1 | - | 6 | 710 | |||
U44426.1 | - | 6 | 504 |