Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vasp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282021.1 | NP_001268950.1 | 3 | 632 | ||
NM_001282022.1 | NP_001268951.1 | 3 | 632 | |||
NM_009499.3 | NP_033525.2 | 3 | 632 | |||
GenBank mRNA | AF084548.1 | - | 3 | 599 | ||
AK140329.1 | - | 3 | 596 | |||
AK150103.1 | - | 3 | 594 | |||
AK155480.1 | - | 3 | 594 | |||
AK157021.1 | - | 3 | 596 | |||
AK170780.1 | - | 3 | 588 | |||
AK171302.1 | - | 3 | 596 | |||
AK171715.1 | - | 3 | 596 | |||
BC010223.1 | - | 3 | 334 | 3500587 | ||
BC015289.1 | - | 3 | 575 | 4240907 |