Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apbb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146085.1 | NP_666197.1 | 8 | 989 | ||
GenBank mRNA | AK141878.1 | - | 8 | 891 | ||
AK171634.1 | - | 7 | 1182 | |||
BC024136.1 | - | 7 | 987 | |||
BC024809.1 | - | 8 | 989 | 5357681 |