Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam178a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081225.1 | NP_001074694.1 | 8 | 2436 | ||
GenBank mRNA | AK028324.1 | - | 4 | 387 | ||
AK030951.1 | - | 4 | 752 | |||
AK031688.1 | - | 8 | 2408 | |||
AK032405.1 | - | 4 | 391 | |||
AK036700.1 | - | 6 | 2013 | |||
AK147588.1 | - | 8 | 2909 | |||
AK161809.1 | - | 4 | 799 | |||
BC060679.1 | - | 4 | 353 | |||
BC150999.1 | - | 8 | 2289 | 9087912 | ||
BC157937.1 | - | 8 | 2289 | 9088117 | ||
BC172097.1 | - | 8 | 2289 | 9054383 |