Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aim1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162970.1 | NP_001156442.1 | 15, 16 | 3988 | ||
GenBank mRNA | AK145690.1 | - | 15, 16 | 3994 | ||
AK155034.1 | - | 3, 4 | 339 | |||
AK165037.1 | - | 2, 3 | 263 | |||
BC042709.1 | - | 10, 11 | 1009 | |||
BC150738.1 | - | 14, 15 | 3917 | 9087649 |