Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172146.2 | NP_742158.1 | 11 | 1973 | ||
GenBank mRNA | AK147750.1 | - | 11 | 1623 | ||
AK148034.1 | - | 11 | 1577 | |||
AK159141.1 | - | 12 | 2123 | |||
AK159859.1 | - | 12 | 2123 | |||
AK160542.1 | - | 11 | 1598 | |||
AK215423.1 | - | 1 | 108 | |||
AK216394.1 | - | 2 | 256 | |||
BC006053.1 | - | 10 | 1815 | |||
BC023841.1 | - | 11 | 1600 | 5346105 |