Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110327.1 | NP_001103797.1 | 16 | 2204 | ||
NM_011806.3 | NP_035936.3 | 17 | 2414 | |||
GenBank mRNA | AK029147.1 | - | 17 | 2356 | ||
AK046201.1 | - | 16 | 2799 | |||
AK078015.1 | - | 16 | 2108 | |||
AK160595.1 | - | 16 | 2177 | |||
BC010272.1 | - | 6 | 774 | |||
BC045141.1 | - | 16 | 2128 | 6307203 | ||
BC060109.1 | - | 17 | 2336 | |||
U70017.1 | - | 17 | 2369 |