Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Arap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178407.3 | NP_848494.2 | 2 | 607 | ||
GenBank mRNA | AK037027.1 | - | 2 | 516 | ||
AK039153.1 | - | 2 | 525 | |||
AK041540.1 | - | 2 | 550 |