Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139220.1 | NP_631966.1 | 2 | 224 | ||
GenBank mRNA | AJ298147.1 | - | 2 | 224 |