Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Adam34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145745.2 | NP_665688.2 | 2 | 296 | ||
GenBank mRNA | AF373288.1 | - | 3 | 380 | ||
AK029590.1 | - | 3 | 349 | |||
BC132102.1 | - | 2 | 250 | 40130379 | ||
BC132292.1 | - | 2 | 250 | 40130569 | ||
BC141203.1 | - | 3 | 339 | 9055815 | ||
BC141204.1 | - | 3 | 342 | 9055816 |