Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146757.1 | NP_666968.1 | 1 | 311 |
Gene Symbol: Olfr566 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011536.1 | NP_001011536.1 | 1 | 302 |
Set Membership: |
![]() |