Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172994.2 | NP_766582.1 | 5, 6 | 871 | ||
GenBank mRNA | AK032023.1 | - | 5, 6 | 774 | ||
AK049830.1 | - | 5, 6 | 757 | |||
AY348866.1 | - | 5, 6 | 873 | |||
BC059811.1 | - | 5, 6 | 871 | 6849535 |