Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201406.1 | NP_958808.1 | 5 | 489 | ||
GenBank mRNA | AK033331.1 | - | 5 | 463 | ||
AK039604.1 | - | 5 | 460 | |||
AK043232.1 | - | 5 | 463 | |||
AK045759.1 | - | 5 | 460 | |||
AK053771.1 | - | 5 | 463 | |||
AK142871.1 | - | 5 | 460 | |||
AK151858.1 | - | 5 | 451 | |||
AK152229.1 | - | 5 | 451 | |||
AK153300.1 | - | 5 | 455 | |||
AK158128.1 | - | 4 | 1087 | |||
AK184954.1 | - | 1 | 36 | |||
AK216471.1 | - | 3 | 260 | |||
AK217146.1 | - | 1 | 23 | |||
BC058979.1 | - | 5 | 489 | 5716747 |