Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ept1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027652.2 | NP_081928.2 | 2 | 249 | ||
GenBank mRNA | AK014530.1 | - | 2 | 195 | ||
AK040494.1 | - | 2 | 228 | |||
AK040745.1 | - | 2 | 158 | |||
AK048011.1 | - | 2 | 198 | |||
AK079325.1 | - | 2 | 225 | |||
AK083973.1 | - | 2 | 225 | |||
AK122544.1 | - | 2 | 165 | |||
AK140416.1 | - | 2 | 221 | |||
BC106097.1 | - | 2 | 218 | 6485810 | ||
BC115783.1 | - | 2 | 188 | 40104916 | ||
BC117551.1 | - | 2 | 183 | 40104922 | ||
BY136287.1 | - | 2 | 249 |