Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081669.2 | NP_001075138.2 | 4 | 594 |
Gene Symbol: Rhox2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029203.2 | NP_083479.1 | 4 | 583 | ||
GenBank mRNA | AK015997.1 | - | 4 | 584 | ||
BC132216.1 | - | 4 | 577 | 40130493 | ||
BC132220.1 | - | 4 | 577 | 40130497 | ||
BC145534.1 | - | 5 | 626 | |||
DQ058639.1 | - | 4 | 584 |
Gene Symbol: Rhox2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114153.1 | NP_001107625.1 | 4 | 599 |
Gene Symbol: Rhox2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100465.1 | NP_001093935.1 | 4 | 583 |