Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177609.1 | NP_001171080.1 | 5 | 1043 | ||
NM_001177610.1 | NP_001171081.1 | 4 | 332 | |||
NM_011665.4 | NP_035795.1 | 4 | 370 | |||
GenBank mRNA | AK003141.1 | - | 4 | 277 | ||
AK005058.1 | - | 4 | 352 | |||
AK011239.1 | - | 4 | 278 | |||
AK012282.1 | - | 4 | 296 | |||
AK088508.1 | - | 4 | 355 | |||
AK150575.1 | - | 4 | 313 | |||
AK160014.1 | - | 4 | 299 | |||
AK160988.1 | - | 4 | 296 | |||
AK165606.1 | - | 4 | 282 | |||
AK165615.1 | - | 4 | 300 | |||
AK166016.1 | - | 4 | 280 | |||
AK166657.1 | - | 5 | 1044 | |||
AK166930.1 | - | 4 | 276 | |||
AK167162.1 | - | 5 | 513 | |||
AK168212.1 | - | 4 | 299 | |||
AK168706.1 | - | 4 | 259 | |||
AK168766.1 | - | 4 | 293 | |||
AK190279.1 | - | 197 | ||||
AK191598.1 | - | 3 | 189 | |||
AK194165.1 | - | 3 | 189 | |||
BC020437.1 | - | 3 | 218 | |||
BC037635.1 | - | 4 | 262 | 4951933 | ||
U31934.1 | - | 3 | 210 | |||
U76416.1 | - | 4 | 306 | |||
U82627.1 | - | 4 | 315 | |||
U94402.1 | - | 4 | 366 | |||
X97575.1 | - | 4 | 247 | |||
X99739.1 | - | 4 | 294 |