Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177609.1 | NP_001171080.1 | 8 | 1262 | ||
NM_001177610.1 | NP_001171081.1 | 7 | 551 | |||
NM_011665.4 | NP_035795.1 | 7 | 589 | |||
GenBank mRNA | AK003141.1 | - | 7 | 496 | ||
AK005058.1 | - | 7 | 571 | |||
AK011239.1 | - | 7 | 497 | |||
AK012282.1 | - | 7 | 515 | |||
AK088508.1 | - | 7 | 574 | |||
AK150575.1 | - | 7 | 532 | |||
AK160014.1 | - | 7 | 518 | |||
AK160988.1 | - | 7 | 515 | |||
AK165606.1 | - | 7 | 501 | |||
AK165615.1 | - | 7 | 519 | |||
AK166016.1 | - | 7 | 499 | |||
AK166657.1 | - | 8 | 1263 | |||
AK166930.1 | - | 7 | 495 | |||
AK167162.1 | - | 8 | 732 | |||
AK168212.1 | - | 7 | 518 | |||
AK168706.1 | - | 7 | 478 | |||
AK168766.1 | - | 7 | 512 | |||
U31934.1 | - | 6 | 429 | |||
U76416.1 | - | 7 | 525 | |||
U82627.1 | - | 7 | 534 | |||
U94402.1 | - | 7 | 585 | |||
X97575.1 | - | 7 | 466 | |||
X99739.1 | - | 7 | 513 |