Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Higd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019814.4 | NP_062788.1 | 4 | 1486 | ||
GenBank mRNA | AB253941.1 | - | 3 | 996 | ||
AK008756.1 | - | 4 | 1113 | |||
AK018360.1 | - | 4 | 1114 | |||
AY028386.1 | - | 4 | 1039 | |||
AY028387.1 | - | 3 | 1545 | |||
BC132371.1 | - | 4 | 1102 | 40130648 | ||
BC151191.1 | - | 4 | 1104 | 8861324 |