Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006353.2 | NP_006344.1 | 2 | 1149 | ||
XM_017010171.1 | XP_016865660.1 | 2 | 1208 | |||
XM_017010172.1 | XP_016865661.1 | 3 | 1272 | |||
XM_017010173.1 | XP_016865662.1 | 4 | 1310 | |||
GenBank mRNA | BC001282.1 | - | 2 | 1093 | 3452950 | |
BC035998.1 | - | 1 | 572 | |||
U90549.1 | - | 1 | 1602 |