Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam229b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183254.1 | NP_899077.1 | 5 | 532 | ||
GenBank mRNA | AK006353.1 | - | 5 | 532 | ||
AK006431.1 | - | 5 | 511 | |||
BC147171.1 | - | 4 | 461 | 9056241 | ||
BC147172.1 | - | 4 | 461 | 9056208 | ||
BC172004.1 | - | 4 | 458 | 9054290 |