Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006361.5 | NP_006352.2 | 2 | 1634 | ||
GenBank mRNA | AY937237.1 | - | 2 | 1643 |