Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006365.2 | NP_006356.1 | 6 | 590 | ||
NR_135264.1 | 5 | 494 | ||||
NR_135268.1 | 6 | 824 | ||||
XM_011509070.1 | XP_011507372.1 | 5 | 903 | |||
XM_011509071.2 | XP_011507373.1 | 5 | 1690 | |||
XM_011509072.1 | XP_011507374.1 | 5 | 513 | |||
XM_011509073.2 | XP_011507375.1 | 4 | 677 | |||
XM_011509074.1 | XP_011507376.1 | 4 | 458 | |||
XM_017000062.1 | XP_016855551.1 | 9 | 1071 | |||
XM_017000063.1 | XP_016855552.1 | 7 | 695 | |||
GenBank mRNA | AK124566.1 | - | 1 | 1845 | ||
AK124870.1 | - | 1 | 668 | |||
BI752228.1 | - | 6 | 813 | |||
DB534652.2 | - | 3 | 269 | |||
HY158402.1 | - | 5 | 372 | |||
U49857.1 | - | 6 | 434 |