Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp474 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025749.3 | NP_080025.3 | 2 | 1031 | ||
GenBank mRNA | AK007266.1 | - | 1 | 83 | ||
AK016749.1 | - | 2 | 1032 | |||
AY350709.1 | - | 4 | 1213 | |||
BC055099.1 | - | 2 | 1027 | 6704066 |