Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other STK25 SIRNA Assays ›
Gene Symbol:
STK25
Gene Name:
serine/threonine kinase 25
Gene Aliases:
SOK1, YSK1
Chromosome Location:
Chr.2: 241494707 - 241509730 on Build GRCh38
UniGene ID:
Hs.516807
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s20569
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: STK25
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001271977.1
NP_001258906.1
8
1161
NM_001271978.1
NP_001258907.1
7
1133
NM_001271979.1
NP_001258908.1
7
930
NM_001271980.1
NP_001258909.1
7
868
NM_001282305.1
NP_001269234.1
7
1128
NM_001282306.1
NP_001269235.1
8
1111
NM_001282307.1
NP_001269236.1
7
1031
NM_001282308.1
NP_001269237.1
7
969
NM_006374.4
NP_006365.2
8
1099
XM_011510493.2
XP_011508795.1
8
1153
XM_011510494.2
XP_011508796.1
8
964
XM_011510495.2
XP_011508797.1
7
1236
XM_011510496.2
XP_011508798.1
8
1153
XM_017003168.1
XP_016858657.1
8
1153
XM_017003169.1
XP_016858658.1
8
1034
XM_017003170.1
XP_016858659.1
7
867
XM_017003171.1
XP_016858660.1
7
954
GenBank mRNA
AK092161.1
-
7
791
AK291808.1
-
7
757
AK291947.1
-
8
988
AK297374.1
-
7
920
AK301214.1
-
8
999
AK303719.1
-
7
1101
AK315966.1
-
7
869
BC007852.2
-
8
1023
4298239
BC015793.1
-
7
1079
4867978
BC091505.1
-
8
1053
30383354
BT019961.1
-
7
808
CR936874.1
-
2
1212
D63780.1
-
7
922
KU178528.1
-
7
808
KU178529.1
-
7
758
KU178530.1
-
6
577
X99325.1
-
7
934
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00179039_m1
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
response to oxidative stress
establishment or maintenance of cell polarity
signal transduction
signal transduction by protein phosphorylation
positive regulation of stress-activated MAPK cascade
intrinsic apoptotic signaling pathway in response to hydrogen peroxide
response to hydrogen peroxide
protein autophosphorylation
positive regulation of axonogenesis
Golgi localization
establishment of Golgi localization
Golgi reassembly
protein kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
protein homodimerization activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0