Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SEMA6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032108.3 | NP_115484.2 | 7, 8 | 835 | ||
XM_011527639.2 | XP_011525941.1 | 7, 8 | 884 | |||
XM_011527640.2 | XP_011525942.1 | 7, 8 | 627 | |||
XM_011527641.1 | XP_011525943.1 | 7, 8 | 884 | |||
GenBank mRNA | AB022433.1 | - | 6, 7 | 611 | ||
AB209278.1 | - | 6, 7 | 597 | |||
AF216389.1 | - | 6, 7 | 576 | |||
AF293363.1 | - | 6, 7 | 576 | |||
AK300037.1 | - | 7, 8 | 835 | |||
AY358939.1 | - | 6, 7 | 591 | |||
BC142617.1 | - | 7, 8 | 778 | 40147342 | ||
FR839673.1 | - | 6, 7 | 573 |