Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023374.3 | NP_075863.2 | 3 | 371 | ||
GenBank mRNA | AK003052.1 | - | 3 | 265 | ||
AK003533.1 | - | 3 | 255 | |||
AK009660.1 | - | 3 | 265 | |||
AK153045.1 | - | 3 | 248 | |||
AK169252.1 | - | 3 | 262 | |||
AK169839.1 | - | 3 | 264 | |||
AK196072.1 | - | 1 | 28 | |||
BC013509.1 | - | 3 | 266 | 4225025 | ||
BC018580.1 | - | 3 | 230 | |||
BC051934.1 | - | 3 | 371 | 5688898 | ||
DQ403000.1 | - | 2 | 162 |