Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fbxo32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026346.3 | NP_080622.1 | 2 | 414 | ||
GenBank mRNA | AF441120.1 | - | 2 | 460 | ||
AK014612.1 | - | 2 | 367 | |||
AK014793.1 | - | 2 | 396 | |||
BC027211.1 | - | 2 | 348 | 3981597 | ||
BU921446.1 | - | 2 | 414 |