Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camsap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081360.1 | NP_001074829.1 | 13 | 3323 | ||
GenBank mRNA | AK028542.1 | - | 11 | 3273 | ||
AK122436.1 | - | 1 | 636 | |||
BC100407.1 | - | 1 | 822 | |||
BC138367.1 | - | 12 | 2899 | 8861387 | ||
BC145258.1 | - | 11 | 2866 | 9053811 |