Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080555.2 | NP_542122.1 | 3 | 797 | ||
GenBank mRNA | AK011276.1 | - | 3 | 796 | ||
AK076183.1 | - | 3 | 493 | |||
AK159136.1 | - | 3 | 883 | |||
AK160056.1 | - | 3 | 697 | |||
BC005558.1 | - | 3 | 784 | 3964817 |