Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024221.3 | NP_077183.1 | 3 | 204 | ||
GenBank mRNA | AK011810.1 | - | 168 | |||
AK014180.1 | - | 3 | 160 | |||
AK014918.1 | - | 3 | 205 | |||
AK084507.1 | - | 2 | 1395 | |||
AK153058.1 | - | 3 | 158 | |||
AK163596.1 | - | 1 | 1481 | |||
AK166631.1 | - | 3 | 206 | |||
AK167798.1 | - | 3 | 161 | |||
BC002188.1 | - | 1 | 28 | |||
BC019512.1 | - | 3 | 177 | 4210204 | ||
BC094468.1 | - | 3 | 150 | 5011266 | ||
CJ159324.1 | - | 3 | 208 |