Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXW10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267585.1 | NP_001254514.1 | 11 | 2159 | ||
NR_051988.1 | 11 | 2805 | ||||
XM_011523625.1 | XP_011521927.1 | 10 | 2312 | |||
XM_011523626.2 | XP_011521928.1 | 10 | 2312 | |||
XM_017024020.1 | XP_016879509.1 | 10 | 2312 | |||
XM_017024021.1 | XP_016879510.1 | 10 | 2312 | |||
GenBank mRNA | AK310383.1 | - | 11 | 2744 | ||
AY729024.1 | - | 11 | 1927 | |||
BC028364.2 | - | 10 | 2233 | 4827344 | ||
BI828401.1 | - | 1 | 41 |