Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026817.3 | NP_081093.2 | 6, 7 | 571 | ||
GenBank mRNA | AK004012.1 | - | 6 | 787 | ||
AK170205.1 | - | 3, 4 | 259 | |||
AK180232.1 | - | 51 | ||||
AK209433.1 | - | 1, 2 | 50 | |||
AK210352.1 | - | 1, 2 | 50 | |||
BC030376.1 | - | 6, 7 | 538 | 5374611 | ||
BQ887131.1 | - | 6, 7 | 563 |