Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301224.1 | NP_001288153.1 | 1, 2 | 380 | ||
NM_006383.3 | NP_006374.1 | 1, 2 | 380 | |||
NR_125435.1 | 1, 2 | 380 | ||||
XM_005254126.3 | XP_005254183.1 | 1, 2 | 821 | |||
XM_006720374.2 | XP_006720437.1 | 1, 2 | 111 | |||
GenBank mRNA | AB012955.1 | - | 1, 2 | 66 | ||
AI817087.1 | - | 1, 2 | 116 | |||
AI857912.1 | - | 1, 2 | 80 | |||
AK293167.1 | - | 1, 2 | 294 | |||
BC047381.1 | - | 1, 2 | 308 | 5287072 | ||
BC057292.1 | - | 1, 2 | 308 | |||
BE042882.1 | - | 1, 2 | 116 | |||
BQ681471.1 | - | 406 | ||||
BT019962.1 | - | 1, 2 | 60 |