Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026912.1 | NP_081188.1 | 7 | 908 | ||
GenBank mRNA | AK005338.1 | - | 2 | 226 | ||
BC016091.1 | - | 7 | 908 | 4503873 |