Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qtnf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204152.1 | NP_001191081.1 | 4 | 1537 | ||
NM_001204153.1 | NP_001191082.1 | 3 | 1492 | |||
NM_028331.5 | NP_082607.3 | 3 | 1703 | |||
GenBank mRNA | AK012868.1 | - | 4 | 1479 | ||
AK051058.1 | - | 3 | 1617 |