Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167750.1 | NP_001161222.1 | 19, 20 | 1878 | ||
NM_001167751.1 | NP_001161223.1 | 19, 20 | 1878 | |||
NM_024260.5 | NP_077222.4 | 19, 20 | 1878 | |||
GenBank mRNA | AK042031.1 | - | 19, 20 | 1858 | ||
AK075898.1 | - | 19, 20 | 1867 | |||
AK129458.1 | - | 19, 20 | 1828 | |||
AK134533.1 | - | 3, 4 | 3677 | |||
AY254697.1 | - | 19, 20 | 1871 | |||
BC002304.1 | - | 4, 5 | 423 | 3591600 | ||
BC036294.1 | - | 19, 20 | 1818 | 3500316 | ||
BC043092.1 | - | 19, 20 | 1835 | 6405356 |