Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Lyrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285959.1 | NP_001272888.1 | 3, 4 | 289 | ||
NM_001285960.1 | NP_001272889.1 | 3, 4 | 422 | |||
NM_029610.3 | NP_083886.1 | 3, 4 | 673 | |||
GenBank mRNA | AK009135.1 | - | 3, 4 | 676 | ||
AK015083.1 | - | 4, 5 | 481 | |||
AK157931.1 | - | 291 | ||||
BC011339.1 | - | 3, 4 | 463 | 3978191 | ||
BY752674.1 | - | 3, 4 | 291 | |||
DV043140.1 | - | 2, 3 | 447 |