Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P3H3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014262.4 | NP_055077.2 | 1, 2 | 563 | ||
GenBank mRNA | AJ430349.1 | - | 2, 3 | 792 | ||
AK025976.1 | - | 1, 2 | 530 | |||
BC017217.2 | - | 1, 2 | 104 | |||
BC067251.1 | - | 1, 2 | 261 | |||
BC080630.1 | - | 1, 2 | 336 |