Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rab11fip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080813.2 | NP_001074282.1 | 4 | 2271 | ||
GenBank mRNA | BC034201.1 | - | 1 | 369 | ||
BC091001.1 | - | 3 | 1710 |